Learn More
Cayman Chemical Stattic, Purity: ≥98%, CAS: 19983-44-9, 25 mg
SDP
Catalog No. 502969529
Encompass_Preferred
A selective inhibitor of STAT3 activation, dimerization and nuclear translocation (IC50 = 5.1 μM at 37°C); 10 μM induces apoptosis in STAT3-dependent breast cancer cells
Catalog No. 50-296-9529 Supplier Cayman Chemical Supplier No. 1459025 MG
A selective inhibitor of STAT3 activation, dimerization and nuclear translocation (IC50 = 5.1 μM at 37°C); 10 μM induces apoptosis in STAT3-dependent breast cancer cells