Learn More
Boca Scientific Inc Antibody Stabilizer TRIS Base, 50ml
Catalog No. NC1443793
Encompass
Antibody Stabilizer conserves the structure of proteins and antibodies to prevent loss of functionality due to storage. With refrigerated storage, Antibody Stabilizer prevents the need for freezing and thawing. Both TRIS-based and PBS-based Antibody Stabilizers are available, allowing the user to select the optimal buffer base for the assay.
This item is not returnable. View return policy
Catalog No. NC1443793 Supplier Boca Scientific Inc Supplier No. 130 050
Antibody Stabilizer conserves the structure of proteins and antibodies to prevent loss of functionality due to storage. With refrigerated storage, Antibody Stabilizer prevents the need for freezing and thawing. Both TRIS-based and PBS-based Antibody Stabilizers are available, allowing the user to select the optimal buffer base for the assay.