Learn More
Cayman Chemical STM2457, Purity: ≥98%, CAS: 2499663-01-1, 25 mg
SDP
Catalog No. 502948762
Encompass_Preferred
An inhibitor of the METTL3/14 complex (IC50 = 16.9 µM); inhibits growth in a panel of AML cell lines (IC50s = 0.6-10.3 µM); inhibits the proliferation, migration, and invasion of HuCC-T1 and HCCC-9810 ICC cells; increases survival in a PDX mouse model of AML at 50 mg/kg
Catalog No. 50-294-8762 Supplier Cayman Chemical Supplier No. 3428025 MG
An inhibitor of the METTL3/14 complex (IC50 = 16.9 µM); inhibits growth in a panel of AML cell lines (IC50s = 0.6-10.3 µM); inhibits the proliferation, migration, and invasion of HuCC-T1 and HCCC-9810 ICC cells; increases survival in a PDX mouse model of AML at 50 mg/kg