Learn More
Cayman Chemical STO-609, Purity: ≥95% (mixture of isomers), CAS: 52029-86-4, 1 mg
SDP
Catalog No. 502990725
Encompass_Preferred
A CaMKK inhibitor (IC50s = 120 and 40 ng/ml for CaMKKα and CaMKKβ, respectively); selective for CaMKKs over CaMKI, CaMKII, CaMKIV, MLCK, PKC, PKA, and p42 MAPK (IC50s = ≥ 10,000 ng/ml for all); inhibits phosphorylation of CaMKI and AMPK in chicken sperm in the presence of extracellular calcium, as well as reduces chicken sperm motility; decreases Npy and Agrp expression and cumulative food intake in mice at 20 μM 0.5 μL/hour, i.c.v.; increases cortical, striatal, and total infarct volume in a mouse model of MCAO-induced focal transient cerebral ischemia at 3 μg/animal, i.c.v.
Catalog No. 50-299-0725 Supplier Cayman Chemical Supplier No. 313581 MG
A CaMKK inhibitor (IC50s = 120 and 40 ng/ml for CaMKKα and CaMKKβ, respectively); selective for CaMKKs over CaMKI, CaMKII, CaMKIV, MLCK, PKC, PKA, and p42 MAPK (IC50s = ≥ 10,000 ng/ml for all); inhibits phosphorylation of CaMKI and AMPK in chicken sperm in the presence of extracellular calcium, as well as reduces chicken sperm motility; decreases Npy and Agrp expression and cumulative food intake in mice at 20 μM 0.5 μL/hour, i.c.v.; increases cortical, striatal, and total infarct volume in a mouse model of MCAO-induced focal transient cerebral ischemia at 3 μg/animal, i.c.v.