Learn More
Cayman Chemical STY-BODIPY, Purity: ≥95%, CAS: 2383063-37-2, 5 mg
SDP
Catalog No. 502940458
Encompass_Preferred
A fluorogenic probe for radical-trapping antioxidant activity; co-autoxidation with a hydrocarbon co-substrate can be quantified by monitoring the loss of absorbance at 571 nm; has been used to measure the activity of radical-trapping antioxidants, as well as the kinetics and stoichiometry of RTA reactions in cell-free assays
Catalog No. 50-294-0458 Supplier Cayman Chemical Supplier No. 270895 MG
A fluorogenic probe for radical-trapping antioxidant activity; co-autoxidation with a hydrocarbon co-substrate can be quantified by monitoring the loss of absorbance at 571 nm; has been used to measure the activity of radical-trapping antioxidants, as well as the kinetics and stoichiometry of RTA reactions in cell-free assays