Learn More
Cayman Chemical SU 16f, Purity: ≥98%, CAS: 251356-45-3, 5 mg
SDP
Catalog No. 502981154
Encompass_Preferred
A potent inhibitor of PDGFRβ (IC50 = 10 nM); inhibits VEGFR2 (IC50 = 140 nM); selectively inhibits PDGF- over VEFG-, FGF-, and EGF-induced cell proliferation (IC50s = 0.11, 10, 10, and 21.9 μM, respectively); accelerates downregulation of fibroblast genes and increases the yield of beating clusters in HFFs treated with 15 compounds to induce a cardiac myocyte-like phenotype
Catalog No. 50-298-1154 Supplier Cayman Chemical Supplier No. 195555 MG
A potent inhibitor of PDGFRβ (IC50 = 10 nM); inhibits VEGFR2 (IC50 = 140 nM); selectively inhibits PDGF- over VEFG-, FGF-, and EGF-induced cell proliferation (IC50s = 0.11, 10, 10, and 21.9 μM, respectively); accelerates downregulation of fibroblast genes and increases the yield of beating clusters in HFFs treated with 15 compounds to induce a cardiac myocyte-like phenotype