Learn More
Cayman Chemical SU 4942, Purity: ≥98%, CAS: 76086-99-2, 500 mg
SDP
Catalog No. 502950600
Encompass_Preferred
A modulator of tyrosine kinase signaling; inhibits VEGF- or acidic FGF-induced mitogenesis of endothelial cells in vitro
Catalog No. 50-295-0600 Supplier Cayman Chemical Supplier No. 32967500 MG
A modulator of tyrosine kinase signaling; inhibits VEGF- or acidic FGF-induced mitogenesis of endothelial cells in vitro