Learn More
Cayman Chemical ANTIBODY SU 9516 10mg
SDP
Catalog No. 502973334
Encompass_Preferred
A potent inhibitor of several CDKs with selectivity for Cdk2 (IC50s = 22, 40, and 200 nM for Cdk2/cyclin A, Cdk1/cyclin B, and Cdk4/cyclin D1); has anti-proliferative and pro-apoptotic activity in tumor cells
Catalog No. 50-297-3334 Supplier Cayman Chemical Supplier No. 1479610 MG
A potent inhibitor of several CDKs with selectivity for Cdk2 (IC50s = 22, 40, and 200 nM for Cdk2/cyclin A, Cdk1/cyclin B, and Cdk4/cyclin D1); has anti-proliferative and pro-apoptotic activity in tumor cells