Learn More
Cayman Chemical SW155246, Purity: ≥98%, CAS: 420092-79-1, 10 mg
SDP
Catalog No. 502986394
Encompass_Preferred
A DNMT1 inhibitor (IC50 = 1.2 μM); selective for human DNMT1 over murine DNMT3 (IC50 = 38 μM); inhibits HeLa and A549 tumor cell growth in a redox-independent manner (IC50s = 8-20 μM); inhibits genomic methylation in HeLa cells from 10 to 18.6 μM
Catalog No. 50-298-6394 Supplier Cayman Chemical Supplier No. 2186510 MG
A DNMT1 inhibitor (IC50 = 1.2 μM); selective for human DNMT1 over murine DNMT3 (IC50 = 38 μM); inhibits HeLa and A549 tumor cell growth in a redox-independent manner (IC50s = 8-20 μM); inhibits genomic methylation in HeLa cells from 10 to 18.6 μM