Learn More
Cayman Chemical T-1095, Purity: ≥98%, CAS: 209746-59-8, 10 mg
SDP
Catalog No. 502917372
Encompass_Preferred
An SGLT1 and SGLT2 inhibitor (IC50s = 22.8 and 2.3 µM, respectively); dietary administration reduces blood glucose and HbA1c levels in a rat model of streptozotocin-induced diabetes at 0.01% w/w; suppresses the development of hyperglycemia and hypertriglyceridemia and reduces HbA1c levels in the KKAγ mouse model of type 2 diabetes; exacerbates post-infarction cardiac dysfunction in a rat model of left anterior descending coronary artery ligation-induced myocardial infarction
Catalog No. 50-291-7372 Supplier Cayman Chemical Supplier No. 3728410 MG
An SGLT1 and SGLT2 inhibitor (IC50s = 22.8 and 2.3 µM, respectively); dietary administration reduces blood glucose and HbA1c levels in a rat model of streptozotocin-induced diabetes at 0.01% w/w; suppresses the development of hyperglycemia and hypertriglyceridemia and reduces HbA1c levels in the KKAγ mouse model of type 2 diabetes; exacerbates post-infarction cardiac dysfunction in a rat model of left anterior descending coronary artery ligation-induced myocardial infarction