Learn More
Cayman Chemical TA-01, Purity: ≥98%, CAS: 1784751-18-3, 1 mg
SDP
Catalog No. 502935728
Encompass_Preferred
An inducer of cardiomyocyte differentiation; increases expression of the cardiomyocyte marker NKX2-5 by 2.2-fold and decreases expression of mesoderm markers and the pre-cardiac marker Isl-1 in HES-3 NKX2-5eGFP/w cells at <5 μM; inhibits CK1ε and CK1Δ and reduces expression of Wnt/β-catenin signaling pathway members
Catalog No. 50-293-5728 Supplier Cayman Chemical Supplier No. 246671 MG
An inducer of cardiomyocyte differentiation; increases expression of the cardiomyocyte marker NKX2-5 by 2.2-fold and decreases expression of mesoderm markers and the pre-cardiac marker Isl-1 in HES-3 NKX2-5eGFP/w cells at <5 μM; inhibits CK1ε and CK1Δ and reduces expression of Wnt/β-catenin signaling pathway members