Learn More
Cayman Chemical TA-02, Purity: ≥98%, CAS: 1784751-19-4, 5 mg
SDP
Catalog No. 502935745
Encompass_Preferred
An inducer of cardiomyocyte differentiation; increases expression of the cardiomyocyte marker NKX2-5 by >2-fold and decreases expression of mesoderm markers and the pre-cardiac marker Isl-1 in HES-3 NKX2-5eGFP/w cells at 5 μM; inhibits CK1ε and CK1D and reduces expression of Wnt/β-catenin signaling pathway members
Catalog No. 50-293-5745 Supplier Cayman Chemical Supplier No. 246685 MG
An inducer of cardiomyocyte differentiation; increases expression of the cardiomyocyte marker NKX2-5 by >2-fold and decreases expression of mesoderm markers and the pre-cardiac marker Isl-1 in HES-3 NKX2-5eGFP/w cells at 5 μM; inhibits CK1ε and CK1D and reduces expression of Wnt/β-catenin signaling pathway members