Learn More
Cayman Chemical TAE684, Purity: ≥95%, CAS: 761439-42-3, 10 mg
SDP
Catalog No. 502964874
Encompass_Preferred
A selective ALK inhibitor that blocks the proliferation of ALCL-derived and ALK-dependent cell lines (IC50s = 2-5 nM); suppresses tumor growth in in vivo models of ALK-positive ALCL and neuroblastoma; also inhibits the activity of the Parkinson’s disease-linked LRRK2 (IC50s = 7.8 and 6.1 nM for wild-type and G2019S mutant LRRK2, respectively)
Catalog No. 50-296-4874 Supplier Cayman Chemical Supplier No. 1767010 MG
A selective ALK inhibitor that blocks the proliferation of ALCL-derived and ALK-dependent cell lines (IC50s = 2-5 nM); suppresses tumor growth in in vivo models of ALK-positive ALCL and neuroblastoma; also inhibits the activity of the Parkinson’s disease-linked LRRK2 (IC50s = 7.8 and 6.1 nM for wild-type and G2019S mutant LRRK2, respectively)