Learn More
Cayman Chemical TAK-733, Purity: ≥98%, CAS: 1035555-63-5, 10 mg
SDP
Catalog No. 502965872
Encompass_Preferred
A MEK1 inhibitor (IC50 = 3.2 nM); inhibits ERK phosphorylation in vitro (EC50 = 1.9 nM); inhibits proliferation of A375 and COLO 205 cells (EC50s = 3.1 and 2.1 nM, respectively) and 14 cutaneous melanoma cell lines (IC50s = V600E mutations
Catalog No. 50-296-5872 Supplier Cayman Chemical Supplier No. 1699810 MG
A MEK1 inhibitor (IC50 = 3.2 nM); inhibits ERK phosphorylation in vitro (EC50 = 1.9 nM); inhibits proliferation of A375 and COLO 205 cells (EC50s = 3.1 and 2.1 nM, respectively) and 14 cutaneous melanoma cell lines (IC50s = V600E mutations