Learn More
Cayman Chemical TAK-981, Purity: ≥95% (mixture of isomers), CAS: 1858276-04-6, 1 mg
SDP
Catalog No. 502947302
Encompass_Preferred
An inhibitor of SAE (IC50 = in vitro; decreases intratumor levels of SUMOlyated proteins and reduces tumor growth in an A20 syngeneic lymphoma mouse model
Catalog No. 50-294-7302 Supplier Cayman Chemical Supplier No. 327411 MG
An inhibitor of SAE (IC50 = in vitro; decreases intratumor levels of SUMOlyated proteins and reduces tumor growth in an A20 syngeneic lymphoma mouse model