Learn More
Cayman Chemical TAS 120, Purity: ≥98%, CAS: 1448169-71-8, 10 mg
SDP
Catalog No. 502984002
Encompass_Preferred
An orally bioavailable inhibitor of FGFRs that irreversibly inhibits all four FGFR subtypes at nanomolar concentrations; blocks the proliferation of human cancer cell lines with FGFR gene abnormalities
Catalog No. 50-298-4002 Supplier Cayman Chemical Supplier No. 2113610 MG
An orally bioavailable inhibitor of FGFRs that irreversibly inhibits all four FGFR subtypes at nanomolar concentrations; blocks the proliferation of human cancer cell lines with FGFR gene abnormalities