Learn More
Cayman Chemical Tazarotene, Purity: ≥98%, CAS: 118292-40-3, 10 mg
SDP
Catalog No. 502994585
Encompass_Preferred
A retinoid prodrug of tazarotenic acid; inhibits proliferation and collagen synthesis by human dermal fibroblasts (1 μM); downregulates markers of keratinocyte differentiation, keratinocyte proliferation, and inflammation and upregulates TIG1 and TIG2 in human-derived psoriatic lesions following topical administration; induces complete reduction of skin utricle diameter in the Rhino mouse (0.1% per day, topical); decreases basal cell carcinoma number and size in a Ptch1+/- mouse model of ultraviolet- or ionizing radiation-induced skin cancer when (0.1% per day, topical)
Catalog No. 50-299-4585 Supplier Cayman Chemical Supplier No. 2355910 MG
A retinoid prodrug of tazarotenic acid; inhibits proliferation and collagen synthesis by human dermal fibroblasts (1 μM); downregulates markers of keratinocyte differentiation, keratinocyte proliferation, and inflammation and upregulates TIG1 and TIG2 in human-derived psoriatic lesions following topical administration; induces complete reduction of skin utricle diameter in the Rhino mouse (0.1% per day, topical); decreases basal cell carcinoma number and size in a Ptch1+/- mouse model of ultraviolet- or ionizing radiation-induced skin cancer when (0.1% per day, topical)