Learn More
Cayman Chemical ANTIBODY TC 13172 500ug
SDP
Catalog No. 502989005
Encompass_Preferred
An inhibitor of MLKL and necroptosis; selective for MLKL over RIPK1 and RIPK3 at 10 µM; inhibits TSZ-induced necroptosis in HT-29 cells (EC50 = 2 nM); inhibits TSZ-induced oligomerization of MLKL and translocation of MLKL to the plasma membrane in HT-29 cells at 100 nM
Catalog No. 50-298-9005 Supplier Cayman Chemical Supplier No. 31034500 UG
An inhibitor of MLKL and necroptosis; selective for MLKL over RIPK1 and RIPK3 at 10 µM; inhibits TSZ-induced necroptosis in HT-29 cells (EC50 = 2 nM); inhibits TSZ-induced oligomerization of MLKL and translocation of MLKL to the plasma membrane in HT-29 cells at 100 nM