Learn More
Cayman Chemical TC-E 5003, Purity: ≥98%, CAS: 17328-16-4, 50 mg
SDP
Catalog No. 502980089
Encompass_Preferred
A PRMT1 inhibitor (IC50 = 1.5 µM); selective over CARM1/PRMT4 and SET7/9 methyltransferases (50s = 1.97 and 4.49 µM, respectively); decreases androgen-dependent gene transcription in vitro; eradicates P. berghei in mice
Catalog No. 50-298-0089 Supplier Cayman Chemical Supplier No. 1771850 MG
A PRMT1 inhibitor (IC50 = 1.5 µM); selective over CARM1/PRMT4 and SET7/9 methyltransferases (50s = 1.97 and 4.49 µM, respectively); decreases androgen-dependent gene transcription in vitro; eradicates P. berghei in mice