Learn More
Cayman Chemical TCS 401, Purity: ≥95%, CAS: 243966-09-8, 25 mg
SDP
Catalog No. 502984108
Encompass_Preferred
A selective inhibitor of PTP1B (Ki = 0.29 µM)
Catalog No. 50-298-4108 Supplier Cayman Chemical Supplier No. 2039325 MG
A selective inhibitor of PTP1B (Ki = 0.29 µM)