Learn More
Cayman Chemical TCS 46b, Purity: ≥98%, CAS: 302799-86-6, 25 mg
SDP
Catalog No. 502945747
Encompass_Preferred
An antagonist of NR1A/2B subunit-containing NMDA receptors (IC50 = 0.0053 µM); selective for NMDA receptors containing NR1A/2B subunits over NMDA receptors containing NR1A/2A and NR1A/2C subunits (IC50s = 35 and >100 µM, respectively); inhibits L-DOPA-induced contralateral rotations in a rat model of 6-OHDA-induced Parkinson's disease at 10 mg/kg; has been radiolabeled and used as an NR1A/2B-selective PET radiotracer
Catalog No. 50-294-5747 Supplier Cayman Chemical Supplier No. 4212325 MG
An antagonist of NR1A/2B subunit-containing NMDA receptors (IC50 = 0.0053 µM); selective for NMDA receptors containing NR1A/2B subunits over NMDA receptors containing NR1A/2A and NR1A/2C subunits (IC50s = 35 and >100 µM, respectively); inhibits L-DOPA-induced contralateral rotations in a rat model of 6-OHDA-induced Parkinson's disease at 10 mg/kg; has been radiolabeled and used as an NR1A/2B-selective PET radiotracer