Learn More
Cayman Chemical TD52, Purity: ≥98%, CAS: 1798328-24-1, 10 mg
SDP
Catalog No. 502917700
Encompass_Preferred
A derivative of erlotinib; decreases the viability of HA22T, Hep3B, PLC/PRF/5, and SK-HEP-1 cells (IC50s = 0.9, 0.9, 0.8, and 1.2 µM, respectively); induces apoptosis in a variety of cancer cells, including HA22T, Hep3B, HCC1937, and MDA-MB-231 cells in a concentration-dependent manner; increases intratumoral PP2A activity, reduces intratumoral CIP2A and phosphorylated Akt levels, and reduces tumor growth in a PLC/PRF/5 mouse xenograft model
Catalog No. 50-291-7700 Supplier Cayman Chemical Supplier No. 3774410 MG
A derivative of erlotinib; decreases the viability of HA22T, Hep3B, PLC/PRF/5, and SK-HEP-1 cells (IC50s = 0.9, 0.9, 0.8, and 1.2 µM, respectively); induces apoptosis in a variety of cancer cells, including HA22T, Hep3B, HCC1937, and MDA-MB-231 cells in a concentration-dependent manner; increases intratumoral PP2A activity, reduces intratumoral CIP2A and phosphorylated Akt levels, and reduces tumor growth in a PLC/PRF/5 mouse xenograft model