Learn More
Cayman Chemical Tegobuvir, Purity: ≥98%, CAS: 1000787-75-6, 1 mg
SDP
Catalog No. 502913905
Encompass_Preferred
A non-nucleoside inhibitor of HCV NS5B; reduces viral infectivity in cells expressing wild-type HCV 1b NS5B but not mutant NS5Bs; selectively inhibits replication of HCV genotypes 1a and 1b over genotype 2a in Huh7-Lunet cells (EC50s = 13.8, 0.8, and 21,900 nM, respectively)
Catalog No. 50-291-3905 Supplier Cayman Chemical Supplier No. 366031 MG
A non-nucleoside inhibitor of HCV NS5B; reduces viral infectivity in cells expressing wild-type HCV 1b NS5B but not mutant NS5Bs; selectively inhibits replication of HCV genotypes 1a and 1b over genotype 2a in Huh7-Lunet cells (EC50s = 13.8, 0.8, and 21,900 nM, respectively)