Learn More
Cayman Chemical Terpineol, Purity: ≥90% (mixture of isomers), CAS: 8000-41-7, 10 g
SDP
Catalog No. 502987547
Encompass_Preferred
A monoterpene alcohol with diverse biological activities; this product is a mixture of α-, β-, and γ-terpineol
Catalog No. 50-298-7547 Supplier Cayman Chemical Supplier No. 2317310 G
A monoterpene alcohol with diverse biological activities; this product is a mixture of α-, β-, and γ-terpineol