Learn More
Cayman Chemical Tetracaine, Purity: ≥98%, CAS: 94-24-6, 1 mg
SDP
Catalog No. 502992614
Encompass_Preferred
An analytical reference standard categorized as an anesthetic; has been found as an adulterant in products marketed as male sexual function enhancers; intended for research and forensic applications
Catalog No. 50-299-2614 Supplier Cayman Chemical Supplier No. 300591 MG
An analytical reference standard categorized as an anesthetic; has been found as an adulterant in products marketed as male sexual function enhancers; intended for research and forensic applications