Learn More
Cayman Chemical Tezacaftor, Purity: ≥98%, CAS: 1152311-62-0, 10 mg
SDP
Catalog No. 502986515
Encompass_Preferred
An investigational compound that promotes the maturation of Delta F508 mutants of CFTR
Catalog No. 50-298-6515 Supplier Cayman Chemical Supplier No. 2148010 MG
An investigational compound that promotes the maturation of Delta F508 mutants of CFTR