Learn More
Cayman Chemical Thimerosal, CAS: 54-64-8, 50 g
SDP
Catalog No. 502947633
Encompass_Preferred
A mercurial antimicrobial agent; bacteriostatic against hemolytic streptococci and fungistatic against H. capsulatum, B. dermatitidis, and S. schenckii; induces apoptosis in HCN-1A human cortical neurons and various neuroblastoma cells from 1-250 µM; induces footpad swelling and dorsal skin contact dermatitis in a mouse model of pseudo-allergic reaction at 80 mg/kg, i.p.
Catalog No. 50-294-7633 Supplier Cayman Chemical Supplier No. 3343250 G
A mercurial antimicrobial agent; bacteriostatic against hemolytic streptococci and fungistatic against H. capsulatum, B. dermatitidis, and S. schenckii; induces apoptosis in HCN-1A human cortical neurons and various neuroblastoma cells from 1-250 µM; induces footpad swelling and dorsal skin contact dermatitis in a mouse model of pseudo-allergic reaction at 80 mg/kg, i.p.