Learn More
Cayman Chemical Thio-TEPA, Purity: ≥98%, CAS: 52-24-4, 250 mg
SDP
Catalog No. 502995668
Encompass_Preferred
An alkylating agent with anticancer activity (IC50 = 1.5 μM against P388 murine leukemia cells); alkylates DNA and inhibits thymidine incorporation (IC50 = 2 μM) inducing irreversible DNA damage; crosslinks guanine nucleotides and induces cell cycle arrest via the formation of interstrand cross-links in L1210 and MCF-7 cancer cells in vitro; reduces the number of pulmonary metastases in mice following subcutaneous injection of melanoma cells; decreases the number of postoperative lung metastases when administered at a dose of 10 mg/kg after surgical removal of an S-91 melanoma tumor established via intramuscular injection in mice
Catalog No. 50-299-5668 Supplier Cayman Chemical Supplier No. 23745250 MG
An alkylating agent with anticancer activity (IC50 = 1.5 μM against P388 murine leukemia cells); alkylates DNA and inhibits thymidine incorporation (IC50 = 2 μM) inducing irreversible DNA damage; crosslinks guanine nucleotides and induces cell cycle arrest via the formation of interstrand cross-links in L1210 and MCF-7 cancer cells in vitro; reduces the number of pulmonary metastases in mice following subcutaneous injection of melanoma cells; decreases the number of postoperative lung metastases when administered at a dose of 10 mg/kg after surgical removal of an S-91 melanoma tumor established via intramuscular injection in mice