Learn More
Cayman Chemical ThioGlo1, Purity: ≥95%, CAS: 137350-66-4, 50 mg
SDP
Catalog No. 502951911
Encompass_Preferred
A thiol-reactive fluorescent probe; ex/em = 384/513 nm, respectively; reacts with sulfite to form a fluorescent adduct with similar spectral characteristics, and interference from sulfite during thiol determination must be accounted for using secondary methods; has been used to monitor oxidative stability in beer
Catalog No. 50-295-1911 Supplier Cayman Chemical Supplier No. 1323550 MG
A thiol-reactive fluorescent probe; ex/em = 384/513 nm, respectively; reacts with sulfite to form a fluorescent adduct with similar spectral characteristics, and interference from sulfite during thiol determination must be accounted for using secondary methods; has been used to monitor oxidative stability in beer