Learn More
Cayman Chemical ThioLox, Purity: ≥98%, CAS: 1202193-89-2, 5 mg
SDP
Catalog No. 502917682
Encompass_Preferred
A 15-LO-1 inhibitor (Ki = 3.3 µM); a P. falciparum aspartate carbamoyltransferase inhibitor (IC50 = 3.32 µM); reduces LPS-induced expression of the mRNAs encoding TNF-α, iNos, Il-1β, Il-6, Il-8, and Il-12b in mouse lung slices at 50 µM; inhibits glutamate-induced increases in lipid peroxidation and mitochondrial superoxide formation, as well as cell death, in mouse HT22 cells at 10 µM
Catalog No. 50-291-7682 Supplier Cayman Chemical Supplier No. 389465 MG
A 15-LO-1 inhibitor (Ki = 3.3 µM); a P. falciparum aspartate carbamoyltransferase inhibitor (IC50 = 3.32 µM); reduces LPS-induced expression of the mRNAs encoding TNF-α, iNos, Il-1β, Il-6, Il-8, and Il-12b in mouse lung slices at 50 µM; inhibits glutamate-induced increases in lipid peroxidation and mitochondrial superoxide formation, as well as cell death, in mouse HT22 cells at 10 µM