Learn More
Cayman Chemical Thymidine, Purity: ≥98%, CAS: 50-89-5, 5 g
SDP
Catalog No. 502985098
Encompass_Preferred
A pyrimidine nucleoside; oral administration in combination with deoxycytidine increases lifespan in a Tk2 H126N knock-in mouse model of TK2 deficiency
Catalog No. 50-298-5098 Supplier Cayman Chemical Supplier No. 205195 G
A pyrimidine nucleoside; oral administration in combination with deoxycytidine increases lifespan in a Tk2 H126N knock-in mouse model of TK2 deficiency