Learn More
Cayman Chemical Thymine, Purity: ≥98%, CAS: 65-71-4, 5 g
SDP
Catalog No. 502917035
Encompass_Preferred
A pyrimidine base; forms complementary base pairs with adenine in DNA
Catalog No. 50-291-7035 Supplier Cayman Chemical Supplier No. 387495 G
A pyrimidine base; forms complementary base pairs with adenine in DNA