Learn More
Cayman Chemical TM5441, Purity: ≥98%, CAS: 1190221-43-2, 1 mg
SDP
Catalog No. 502994568
Encompass_Preferred
A PAI-1 inhibitor; decreases survival of HT1080, HCT116, Daoy, MDA-MB-231, and Jurkat cancer cells (IC50s = 13.9-51.1 µM); inhibits branching of HUVECs (50 µM); disrupts tumor vasculature in mouse xenograft models (20 mg/kg per day); prevents L-NAME-induced hypertension and cardiac hypertrophy; protects against NAFLD in mice induced by high-fat diet
Catalog No. 50-299-4568 Supplier Cayman Chemical Supplier No. 229651 MG
A PAI-1 inhibitor; decreases survival of HT1080, HCT116, Daoy, MDA-MB-231, and Jurkat cancer cells (IC50s = 13.9-51.1 µM); inhibits branching of HUVECs (50 µM); disrupts tumor vasculature in mouse xenograft models (20 mg/kg per day); prevents L-NAME-induced hypertension and cardiac hypertrophy; protects against NAFLD in mice induced by high-fat diet