Learn More
Cayman Chemical TTNPB, Purity: ≥98%, CAS: 71441-28-6, 1 mg
SDP
Catalog No. 502971821
Encompass_Preferred
An analog of retinoic acid that potently and selectively activates retinoic acid receptors (EC50s = 21, 4, and 2.4 nM for RARα, RARβ, and RARγ, respectively); used to study RAR action in epidermal cell proliferation, embryogenesis, and stem cell differentiation
Catalog No. 50-297-1821 Supplier Cayman Chemical Supplier No. 161441 MG
An analog of retinoic acid that potently and selectively activates retinoic acid receptors (EC50s = 21, 4, and 2.4 nM for RARα, RARβ, and RARγ, respectively); used to study RAR action in epidermal cell proliferation, embryogenesis, and stem cell differentiation