Learn More
Cayman Chemical TPTZ, Purity: ≥95%, CAS: 3682-35-7, 5 g
SDP
Catalog No. 502939202
Encompass_Preferred
A colorimetric reagent for the detection of iron; forms a complex with Fe(II) that can be quantified by colorimetric detection at 594 nm as a measure of iron concentration
Catalog No. 50-293-9202 Supplier Cayman Chemical Supplier No. 282745 G
A colorimetric reagent for the detection of iron; forms a complex with Fe(II) that can be quantified by colorimetric detection at 594 nm as a measure of iron concentration