Learn More
Cayman Chemical ANTIBODY TPU-0037C 500ug
SDP
Catalog No. 502933381
Encompass_Preferred
A Streptomyces metabolite; structurally similar to lydicamycin; active against Gram-positive bacteria (MICs = 0.39-3.13 UG/ml), including methicillin-resistant S. aureus strains (MIC = 3.13 UG/ml); ineffective against Gram-negative bacteria (MICs = >50 UG/ml)
Catalog No. 50-293-3381 Supplier Cayman Chemical Supplier No. 24179500 UG
A Streptomyces metabolite; structurally similar to lydicamycin; active against Gram-positive bacteria (MICs = 0.39-3.13 UG/ml), including methicillin-resistant S. aureus strains (MIC = 3.13 UG/ml); ineffective against Gram-negative bacteria (MICs = >50 UG/ml)