Learn More
Cayman Chemical TPX-0022, Purity: ≥98%, CAS: 2271119-26-5, 25 mg
SDP
Catalog No. 502911563
Encompass_Preferred
An inhibitor of Src, MET, and FMS kinases (IC50s = 0.12, 0.14, and 0.76 nM, respectively); inhibits the proliferation of MKN45 and SNU-5 human gastric cancer cells (IC50s = 0.2 and 0.17 nM, respectively), as well as Ba/F3 pro-B cells expressing TEL-CSF1R (IC50s = 19.3 nM); inhibits cell migration of HCC827 cells at 100 nM; inhibits tumor growth in a MKN45 mouse xenograft model at 10 and 30 mg/kg; inhibits phosphorylation of MET at Tyr1234 and Tyr1349 in the same model at 10 mg/kg
Catalog No. 50-291-1563 Supplier Cayman Chemical Supplier No. 3611825 MG
An inhibitor of Src, MET, and FMS kinases (IC50s = 0.12, 0.14, and 0.76 nM, respectively); inhibits the proliferation of MKN45 and SNU-5 human gastric cancer cells (IC50s = 0.2 and 0.17 nM, respectively), as well as Ba/F3 pro-B cells expressing TEL-CSF1R (IC50s = 19.3 nM); inhibits cell migration of HCC827 cells at 100 nM; inhibits tumor growth in a MKN45 mouse xenograft model at 10 and 30 mg/kg; inhibits phosphorylation of MET at Tyr1234 and Tyr1349 in the same model at 10 mg/kg