Learn More
Cayman Chemical Torasemide, Purity: ≥98%, CAS: 56211-40-6, 50 mg
SDP
Catalog No. 502981504
Encompass_Preferred
An analytical reference standard categorized as a diuretic; has been abused as a performance-enhancing drug and masking agent in doping in sports; intended for research and forensic applications
Catalog No. 50-298-1504 Supplier Cayman Chemical Supplier No. 2131250 MG
An analytical reference standard categorized as a diuretic; has been abused as a performance-enhancing drug and masking agent in doping in sports; intended for research and forensic applications