Learn More
Cayman Chemical Triflusal, Purity: ≥98%, CAS: 322-79-2, 10 mg
SDP
Catalog No. 502992482
Encompass_Preferred
A platelet aggregation inhibitor (IC50s = 48.3 and 693 µM in isolated human whole blood and platelet-rich plasma, respectively); inhibits arachidonic acid-induced TXB2 and PGF1α production (IC50s = 468 and 339 µM, respectively); decreases IgG-ovalbumin immune complex-induced NF-κB nuclear translocation and NO production in isolated rat peritoneal macrophages at 1,000 µM; reduces infarct volume and peri-infarct levels of OX-6 in a rat model of focal ischemia induced by permanent MCAO at 30 mg/kg; reduces increases in APP levels induced by amyloid-β (25-35) in rats at 30 mg/kg
Catalog No. 50-299-2482 Supplier Cayman Chemical Supplier No. 3027410 MG
A platelet aggregation inhibitor (IC50s = 48.3 and 693 µM in isolated human whole blood and platelet-rich plasma, respectively); inhibits arachidonic acid-induced TXB2 and PGF1α production (IC50s = 468 and 339 µM, respectively); decreases IgG-ovalbumin immune complex-induced NF-κB nuclear translocation and NO production in isolated rat peritoneal macrophages at 1,000 µM; reduces infarct volume and peri-infarct levels of OX-6 in a rat model of focal ischemia induced by permanent MCAO at 30 mg/kg; reduces increases in APP levels induced by amyloid-β (25-35) in rats at 30 mg/kg