Learn More
Cayman Chemical TVB-3166, Purity: ≥98%, CAS: 1533438-83-3, 10 mg
SDP
Catalog No. 502947961
Encompass_Preferred
A FASN inhibitor (IC50 = 0.042 µM); inhibits palmitate biosynthesis in Calu-6 and MRC-5 cells; induces apoptosis in Calu-6 and 22Rv1 cells at 0.2 µM; reduces tumor growth in an OVCAR-8 mouse xenograft model and K-Ras-wild-type or -mutant PDX mouse models at 60 mg/kg; reduces lung viral titers in mice infected with RSV-A at 50 mg/kg
Catalog No. 50-294-7961 Supplier Cayman Chemical Supplier No. 3353310 MG
A FASN inhibitor (IC50 = 0.042 µM); inhibits palmitate biosynthesis in Calu-6 and MRC-5 cells; induces apoptosis in Calu-6 and 22Rv1 cells at 0.2 µM; reduces tumor growth in an OVCAR-8 mouse xenograft model and K-Ras-wild-type or -mutant PDX mouse models at 60 mg/kg; reduces lung viral titers in mice infected with RSV-A at 50 mg/kg