Learn More
Cayman Chemical TW-37, Purity: ≥98%, CAS: 877877-35-5, 25 mg
SDP
Catalog No. 502982100
Encompass_Preferred
An inhibitor of the Bcl-2 family proteins Bcl-2, Mcl-1, and Bcl-xL (Kis = 120, 260, and 1,100 nM, respectively); induces apoptosis, inhibits migration and capillary sprouting in endothelial cells; has apoptotic action against leukemia, lymphoma, and pancreatic cancer cells
Catalog No. 50-298-2100 Supplier Cayman Chemical Supplier No. 2099925 MG
An inhibitor of the Bcl-2 family proteins Bcl-2, Mcl-1, and Bcl-xL (Kis = 120, 260, and 1,100 nM, respectively); induces apoptosis, inhibits migration and capillary sprouting in endothelial cells; has apoptotic action against leukemia, lymphoma, and pancreatic cancer cells