Learn More
Cayman Chemical TX-1918, Purity: ≥98%, CAS: 503473-32-3, 1 mg
SDP
Catalog No. 502985493
Encompass_Preferred
A selective eEF-2K inhibitor (IC50 = 440 nM); disrupts the proliferation of HepG2 and HCT116 tumor cells (EC50s = 2.07 and 230 µM, respectively) with low mitochondrial toxicity
Catalog No. 50-298-5493 Supplier Cayman Chemical Supplier No. 208401 MG
A selective eEF-2K inhibitor (IC50 = 440 nM); disrupts the proliferation of HepG2 and HCT116 tumor cells (EC50s = 2.07 and 230 µM, respectively) with low mitochondrial toxicity