Learn More
Cayman Chemical UK 383367, Purity: ≥98%, CAS: 348622-88-8, 10 mg
SDP
Catalog No. 502973133
Encompass_Preferred
A potent, selective inhibitor of BMP1 (IC50 = 44 nM); effective in a cell-based model of collagen deposition is very effective at penetrating human skin in vitro
Catalog No. 50-297-3133 Supplier Cayman Chemical Supplier No. 1502910 MG
A potent, selective inhibitor of BMP1 (IC50 = 44 nM); effective in a cell-based model of collagen deposition is very effective at penetrating human skin in vitro