Learn More
Cayman Chemical UNC0379, Purity: ≥95%, CAS: 1620401-82-2, 10 mg
SDP
Catalog No. 502970493
Encompass_Preferred
A selective inhibitor of the lysine methyltransferase SET8 (IC50 = 7.3 µM; Kd = 18.3 µM)
Catalog No. 50-297-0493 Supplier Cayman Chemical Supplier No. 1640010 MG
A selective inhibitor of the lysine methyltransferase SET8 (IC50 = 7.3 µM; Kd = 18.3 µM)