Learn More
Cayman Chemical UNC0642, Purity: ≥95%, CAS: 1481677-78-4, 1 mg
SDP
Catalog No. 502970697
Encompass_Preferred
A selective inhibitor of G9a and GLP methyltransferases that competitively inhibits binding of H3K9 substrates with a Ki = 3.7 nM; reduces H3K9 dimethylation levels in MDA-MB-231 and PANC-1 cells (IC50s = 110 and 40 nM, respectively); displays improved pharmacokinetic properties relative to UNC0638
Catalog No. 50-297-0697 Supplier Cayman Chemical Supplier No. 146041 MG
A selective inhibitor of G9a and GLP methyltransferases that competitively inhibits binding of H3K9 substrates with a Ki = 3.7 nM; reduces H3K9 dimethylation levels in MDA-MB-231 and PANC-1 cells (IC50s = 110 and 40 nM, respectively); displays improved pharmacokinetic properties relative to UNC0638