Learn More
Cayman Chemical UNC569, Purity: ≥98%, CAS: 1350547-65-7, 10 mg
SDP
Catalog No. 502994959
Encompass_Preferred
A TAM family kinase inhibitor (IC50s = 2.9, 37, and 48 nM for Mer, Axl, and Tyro3, respectively); antiproliferative against ALL cells (IC50s = 0.5 and 1.2 μM for 697 and Jurkat cell lines, respectively); inhibits Mer phosphorylation (IC50s = 141 and 193 nM in 697 and Jurkat cell lines, respectively); decreases tumor burden by 47.8% relative to controls in human MYC transgenic zebrafish (4 μM); delays leukemia onset, reduces CNS infiltration, and prolongs survival of mice implanted with patient-derived Mer-expressing ALL primary cells (10 mg/kg)
Catalog No. 50-299-4959 Supplier Cayman Chemical Supplier No. 2296610 MG
A TAM family kinase inhibitor (IC50s = 2.9, 37, and 48 nM for Mer, Axl, and Tyro3, respectively); antiproliferative against ALL cells (IC50s = 0.5 and 1.2 μM for 697 and Jurkat cell lines, respectively); inhibits Mer phosphorylation (IC50s = 141 and 193 nM in 697 and Jurkat cell lines, respectively); decreases tumor burden by 47.8% relative to controls in human MYC transgenic zebrafish (4 μM); delays leukemia onset, reduces CNS infiltration, and prolongs survival of mice implanted with patient-derived Mer-expressing ALL primary cells (10 mg/kg)