Learn More
Cayman Chemical UNC926, Purity: ≥98%, CAS: 1184136-10-4, 10 mg
SDP
Catalog No. 502937076
Encompass_Preferred
An antagonist of L3MBTL1, L3MBTL3, and L3MBTL4; inhibits methyl-lysine histone peptide substrate binding of L3MBTL1, L3MBTL3, and L3MBTL4 (IC50s =3.9, 3.2, and 15.6 μM, respectively) selectively over MBTD1, SFMBT1, and CBX7 (IC50s = >30 μM)
Catalog No. 50-293-7076 Supplier Cayman Chemical Supplier No. 2730210 MG
An antagonist of L3MBTL1, L3MBTL3, and L3MBTL4; inhibits methyl-lysine histone peptide substrate binding of L3MBTL1, L3MBTL3, and L3MBTL4 (IC50s =3.9, 3.2, and 15.6 μM, respectively) selectively over MBTD1, SFMBT1, and CBX7 (IC50s = >30 μM)