Learn More
Cayman Chemical ANTIBODY UrodIlatIn 500ug
SDP
Catalog No. 502968190
Encompass_Preferred
A renal natriuretic peptide that regulates sodium and water reabsorption in the kidney; has pharmacological effects on renal, cardiovascular, and pulmonary parameters in animals
Catalog No. 50-296-8190 Supplier Cayman Chemical Supplier No. 17400500 UG
A renal natriuretic peptide that regulates sodium and water reabsorption in the kidney; has pharmacological effects on renal, cardiovascular, and pulmonary parameters in animals