Learn More
Cayman Chemical ANTIBODY USP8-IN-1 5mg
SDP
Catalog No. 502943090
Encompass_Preferred
A USP8 inhibitor (IC50 = 2.6 µM); selective for USP8 over USP7 (IC50 = >20 µM); inhibits colony formation by H1975 cells (IC50 = 30.8 µM)
Catalog No. 50-294-3090 Supplier Cayman Chemical Supplier No. 415985 MG
A USP8 inhibitor (IC50 = 2.6 µM); selective for USP8 over USP7 (IC50 = >20 µM); inhibits colony formation by H1975 cells (IC50 = 30.8 µM)