Learn More
Cayman Chemical USP8-IN-2, Purity: ≥98%, CAS: 2477651-11-7, 5 mg
SDP
Catalog No. 502945729
Encompass_Preferred
A USP8 inhibitor (IC50 = 6 µM)
Catalog No. 50-294-5729 Supplier Cayman Chemical Supplier No. 415995 MG
A USP8 inhibitor (IC50 = 6 µM)